Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMKK2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 115491AP150UL

Description
calmodulin-dependent kinase ; involved in activating Cam-dependent kinase IV.Specifications
CAMKK2 | |
Polyclonal | |
Unconjugated | |
CAMKK2 | |
CaM kinase kinase 2, CaM kinase kinase beta, CaM KK 2, CaM KK beta, CAMKK, CaMKK 2, CaMKK beta, CAMKK2, CAMKKB, KIAA0787 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10645, 207565 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C078, Q96RR4 | |
CAMKK2 | |
CAMKK2 Fusion Protein Ag2093 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction