Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CaMKV Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117829
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5117829 Supplier Invitrogen™ Supplier No. PA5117829
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CaMKV Polyclonal Antibody for Western Blot, ICC/IF, IP

CaMKV does not appear to have detectable kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CaMKV
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene CAMKV
Gene Accession No. Q8NCB2
Gene Alias 1G5; 1G5z; BB074618; BC017634; calcium/calmodulin dependent protein; CaM kinase like vesicle associated; CaM kinase-like vesicle-associated; CaM kinase-like vesicle-associated b; caM kinase-like vesicle-associated protein; CAMK5; Camkv; camkvb; DKFZp564G2082; hypothetical protein MGC63506; testis tissue sperm-binding protein Li 52e; VACAMKL; vesicle-associated calmodulin-binding protein; zgc:63506
Gene Symbols CAMKV
Host Species Rabbit
Immunogen E. coli-derived Human CAMKV fragment.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79012
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.