Learn More
Invitrogen™ CaMKV Polyclonal Antibody

Description
Positive Samples: Mouse eye, Mouse spleen, Rat eye Immunogen sequence: MPFGCVTLGD KKNYNQPSEV TDRYDLGQVI KTEEFCEIFR AKDKTTGKLH TCKKFQKRDG RKVRKAAKNE IGILKMVKHP NILQLVDVFV TRKEYFIFLE LATGREVFDW ILDQGYYSER DTSNVVRQVL EAVAYLHSLK IVHRNLKLEN LVYYNRLKNS KIVISDFHLA KLENGLIKEP CGTPEYLAPE VVGRQRYGRP VDCWAIGVIM YILLSGNPPF YEEVEEDDYE NHDKNLFRKI LAGDYEFDSP YWDDISQAAK DLVTRLMEVE QDQRITAEEA ISHEW.
Specifications
Specifications
| Antigen | CaMKV |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | CAMKV |
| Gene Accession No. | Q3UHL1, Q63092, Q8NCB2 |
| Gene Alias | 1G5; 1G5z; BB074618; BC017634; calcium/calmodulin dependent protein; CaM kinase like vesicle associated; CaM kinase-like vesicle-associated; CaM kinase-like vesicle-associated b; caM kinase-like vesicle-associated protein; CAMK5; Camkv; camkvb; DKFZp564G2082; hypothetical protein MGC63506; testis tissue sperm-binding protein Li 52e; VACAMKL; vesicle-associated calmodulin-binding protein; zgc:63506 |
| Gene Symbols | CAMKV |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.