Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMLG Antibody (OTI1A3), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270331AF750
Description
CAMLG Monoclonal specifically detects CAMLG in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CAMLG | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
CAMLG | |
Full length recombinant protein of human CAMLG (NP_001736) produced in HEK293T cells. | |
Signal Transduction | |
819 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1A3 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
calcium modulating ligand, calcium signal-modulating cyclophilin ligand, calcium-signal modulating cyclophilin ligand, CAMLcalcium-modulating cyclophilin ligand, cyclophilin B-binding protein, MGC163197 | |
Mouse | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction