Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Camostat mesylate
GREENER_CHOICESDP

Catalog No. 319350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-935-0 50 mg
31-931-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 31-935-0 Supplier R&D Systems™ Supplier No. 3193/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Orally active protease inhibitor

Camostat mesylate is an orally active protease inhibitor. Inhibits trypsin and various inflammatory proteases including plasmin, kallikrein and thrombin. Suppresses pancreatitis-induced pain in rats following oral administration. Also inhibits transmembrane serine protease TMPRSS2 and partially blocks entry of SARS-Cov-2 (COVID-19) into lung cells in vitro.

R&D Systems products are for biomedical research use only. They are not intended for human or veterinary use.

Chemical Identifiers

CAS 59721-29-8
Molecular Formula C21H26N4O8S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 494.52
Purity >98%
Molecular Weight (g/mol) 494.52
Chemical Name or Material Camostat mesylate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.