Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMP/Cathelicidin Antibody (OSX12), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP146781AF350
Description
CAMP/Cathelicidin Monoclonal specifically detects CAMP/Cathelicidin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CAMP/Cathelicidin | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CAMP | |
A synthetic peptide from human CAMP/Cathelicidin conjugated to an immunogenic carrier protein has been used as the antigen. | |
0.1 mL | |
820 | |
Human | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OSX12 | |
Western Blot 5-10 ug/ml, Flow Cytometry 5-10 ug/ml, Immunohistochemistry 5-10 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5-10 ug/ml | |
18 kDa cationic antimicrobial protein, Antibacterial peptide FALL-39, CAP18, CAP-18, cathelicidin antimicrobial peptide, CRAMP, epididymis secretory sperm binding protein, FALL39, FALL-39, FALL-39 peptide antibiotic, hCAP-181, HSD26, LL37 | |
Mouse | |
Ammonium sulfate precipitation | |
Primary | |
Specific for Cathelicidin antimicrobial peptide. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction