Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva cAMP Direct BioTrak™ EIA

Catalog No. 45001770 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 96 Determinations
96 Determinations
Assay Sensitivity:
38.4 pg/mL (non-Ac)
38.4 pg/mL (non-Ac), 14 pg/mL (Ac)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Assay Sensitivity
45-001-770 5 x 96 Determinations 38.4 pg/mL (non-Ac), 14 pg/mL (Ac)
45-001-769 96 Determinations 38.4 pg/mL (non-Ac)
45-001-768 96 Determinations 38.4 pg/mL (non-Ac), 14 pg/mL (Ac)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45-001-770 Supplier Cytiva Supplier No. RPN2255

Use for direct measurement of cAMP in cell lysates without any need for removal of extraction reagents. Cytiva Life Sciences™ cAMP Direct BioTrak™ EIA represents a convenient, high-sensitivity, peroxidase-based technology that enables the amount of protein present in a sample to be measured in a single day.

  • Flexible method-choice of assay protocols
  • Rapid assay protocol
  • Non-radioactive
  • Precise and accurate measurement
  • Ready-to-use substrate
  • Color coded reagents
  • Five-hour protocol

  • Kits available with or without acetylation reagents
  • Bulk kit provided without lysis buffers
  • Dual range: 25–6400 fmol/well [non-acetylation], 2–128 fmol/well [acetylation]
  • Sensitivity: 38.4pg/mL [non-acetylation] and 14pg/mL [acetylation]

TRUSTED_SUSTAINABILITY

Specifications

Assay Sensitivity 38.4 pg/mL (non-Ac), 14 pg/mL (Ac)
Product Type cAMP biotrak Enzyme immunoassay (EIA) system
Sufficient For 5 x 96-well Microplate
For Use With (Application) Used for research purposes, eliminates lengthy sample extraction procedures and allow direct cAMP measurement
Kit Contents 96-well coated plates, Assay buffer, Non-acetylation assay standard, Peroxidase conjugate antiserum, TMB substrate, Wash buffer
Quantity 5 x 96 Determinations
Storage Requirements 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.