Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMP/LL37/FALL39/Cathelicidin Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176864AF700
Description
CAMP/LL37/FALL39/Cathelicidin Polyclonal specifically detects CAMP/LL37/FALL39/Cathelicidin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CAMP/LL37/FALL39/Cathelicidin | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
18 kDa cationic antimicrobial protein, CAP18CRAMP, cathelicidin antimicrobial peptide, FALL-39, FALL39CAP-18, hCAP-18, LL37 | |
Rabbit | |
Affinity Purified | |
Immunology | |
820.0 | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CAMP | |
Antibody was raised against a 17 amino acid synthetic peptide from an internal portion of the human Cathelicidin protein. The immunogen is located within amino acids 50 - 100 of Cathelicidin. | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction