Learn More
cAMP Monoclonal Antibody (4H2B6), THE™, GenScript™
Mouse Monoclonal Antibody
Supplier: Genscript Corporation A01509200

Description
Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. Adenosine 3, 5-cyclic monophosphate (cyclic AMP; cAMP) is one of the most important intracellular secondary messengers for transduction events. cAMP is also involved in regulating neuronal, glandular, cardiovascular, immune, and other functions and actions. A number of hormones, such as ACTH, TSH, FSH, and LH, are known to activate cAMP through the action of the enzyme adenylate cyclase, which converts ATP to cAMP. There remains considerable interest in the measurement of intracellular cAMP in tissues and cell cultures, and this may help to provide an understanding of the physiology and pathology of many disease states. Due to the involvement of cAMP in amplifying the response of ligand binding, the second messenger cAMP has been largely employed to monitor the activation of GPCR to facilitate therapeutic drug discovery.GenScript THE™ cAMP Antibody, mAb, Mouse is developed using 3, 5-cyclic AMP-8-KLH as immunogen.
cAMP Monoclonal antibody specifically detects cAMP in Chemical samples. It is validated for ELISASpecifications
cAMP | |
Monoclonal | |
Unconjugated | |
18 kDa cationic antimicrobial protein; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; CAMP; CAP18; CAP-18; Cathelicidin antimicrobial peptide; cathelin-like protein; Cathelin-related antimicrobial peptide; CLP; Cnlp; Cramp; Cyclic AMP; FALL39; FALL-39; FALL-39 peptide antibiotic; HSD26; LL37; MCLP | |
3', 5'-cyclic AMP-8-KLH | |
200 μg | |
Primary | |
Antibody | |
IgG2a κ |
ELISA | |
4H2B6 | |
PBS with 0.02% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Chemical |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.