Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ cAMP Monoclonal Antibody (9H4C4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515393
Description
MA5-15393 targets cAMP in indirect ELISA applications. The MA5-15393 immunogen is cAMP conjugated to KLH.
Cyclic adenosine monophosphate (cAMP) plays a key role as an intracellular second messenger for transduction events that follow a number of extracellular signals. The G-Protein Coupled Receptors (GPCR) is the largest family of cell surface receptors. They can be activated by different ligands, such as neurotransmitters, hormones, ions, small molecules, peptides, and other physiological signaling molecules. Typically, the binding of the ligands to its receptor resulting in the activation of G-proteins, in return, activates the effector adenylyl cyclase evoking the production of cAMP. The activation of a protein kinase by cAMP results in the phosphorylation of substrate proteins. Currently successful drugs in marketing have been developed to target these receptors. Among the GPCRs, ~367 receptors are potential drug development targets, but only about 20 have been used to generate therapeutically and commercially successful drugs so far. Because the involvement of cAMP can amplify the response of the ligand binding, the second messenger cAMP has been largely employed to monitor the activation of the GPCR to facilitate the therapeutic drug discovery.
Specifications
cAMP | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Mouse | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Chemical |
ELISA | |
9H4C4 | |
Unconjugated | |
18 kDa cationic antimicrobial protein; Antibacterial peptide FALL-39; Antibacterial peptide LL-37; CAMP; CAP18; CAP-18; Cathelicidin antimicrobial peptide; cathelin-like protein; Cathelin-related antimicrobial peptide; CLP; Cnlp; Cramp; Cyclic AMP; FALL39; FALL-39; FALL-39 peptide antibiotic; HSD26; LL37; MCLP | |
cAMP, conjugated to KLH. | |
RUO | |
Chemical | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction