Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Camphanic Chloride 97.0+%, TCI America™
SDP

Catalog No. C102225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C102225G 25 g
C10221G 1 g
C10225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C102225G Supplier TCI America Supplier No. C102225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39637-74-6
Molecular Formula C10H13ClO3
Molecular Weight (g/mol) 216.661
MDL Number MFCD00135626
InChI Key PAXWODJTHKJQDZ-QVDQXJPCSA-N
Synonym 1s-4,7,7-trimethyl-3-oxo-2-oxabicyclo 2.2.1 heptane-1-carbonyl chloride, --1s,4r-camphanoyl chloride, 1s---camphanic acid chloride
PubChem CID 12364938
IUPAC Name (4S)-1,7,7-trimethyl-2-oxo-3-oxabicyclo[2.2.1]heptane-4-carbonyl chloride
SMILES CC1(C2(CCC1(OC2=O)C(=O)Cl)C)C

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 216.66
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Camphanic Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.