Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Camphor
SCI_ED

Catalog No. s25232 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Lab Grade
Reagent Grade
This item is not returnable. View return policy
This item is not returnable. View return policy
Kill and preserve moths

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 76-22-2
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.24
MDL Number MFCD00074738,MFCD00064149
InChI Key DSSYKIVIOFKYAU-UHFFFAOYNA-N
Synonym camphor, dl-camphor, 2-camphanone, 2-bornanone, l---camphor, +/--camphor, root bark oil, 1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, bornan-2-one, alphanon
PubChem CID 2537
ChEBI CHEBI:36773
IUPAC Name 1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
SMILES CC1(C)C2CCC1(C)C(=O)C2

Specifications

Color White
Density 0.992g/mL at 25°C
Assay Percent Range >96% w/v
Packaging Lab Grade
Linear Formula C10H16O
Quantity 25 g
Formula Weight 152.25
Chemical Name or Material Camphor
ShelfLife 5 years
DOTInformation DOT Class 4.1, III, : Flammable Solid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.