Alfa Aesar™ (+/-)-Camphor, 96%

Manufacturer: Alfa Aesar™ A1093622

Quantity 100g

 View more versions of this product

Catalog No. AAA1093622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 76-22-2
    Molecular Formula C10H16O
    Formula Weight 152.24
    MDL Number MFCD00074738
    UN Number UN2717
    Beilstein 1907611
    Merck Index 14,1732
    Synonym DL-2-Bornanone
    Chemical Name or Material (±)-Camphor

    Specifications

    Formula Weight 152.24
    Assay Percent Range 96%
    Health Hazard 3 GHS P Statement
    P210-P280g-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Health Hazard 2 GHS H Statement
    H228-H315-H319
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.
    UN Number UN2717
    Beilstein 1907611
    Solubility Information Soluble in alcohols, ether, chloroform, benzene
    RTECS Number EX1225000
    Flash Point 64°C (147°F)
    Odor Camphor-like
    Molecular Formula C10H16O
    MDL Number MFCD00074738
    Synonym DL-2-Bornanone
    TSCA TSCA
    Merck Index 14,1732
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: CAMPHOR
    Boiling Point 204°C
    CAS 76-22-2
    Melting Point 172° to 176°C
    Quantity 100g
    Chemical Name or Material (±)-Camphor
    Assay 96%

    Description

    Applications
    Camphor is used in the preparation of mothballs. It acts as a plasticizer for nitrocellulose, and an ingredient for fireworks and explosive munitions. It is useful in the treatment of sprains, swellings and inflammation. It is also used to synthesize carbon nanotubes by chemical vapor deposition process of camphor.

    Solubility
    Soluble in acetone, ethanol, diethylether, chloroform and acetic acid.

    Notes
    Incompatible with strong oxidizing agents, strong reducing agents and chlorinated solvents.

    GHS H Statement
    H228-H315-H319
    Flammable solid.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P210-P280g-P305+P351+P338
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit