Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Camphor, Synthetic, USP, Spectrum™ Chemical
SDP

Catalog Number 18601407
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
110 lb.
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
76-22-2
C10H16O
152.24
MFCD00074738,MFCD00064149
DSSYKIVIOFKYAU-UHFFFAOYNA-N
1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
CC1(C)C2CCC1(C)C(=O)C2
76-22-2
C10H16O
152.24
MFCD00074738,MFCD00064149
DSSYKIVIOFKYAU-UHFFFAOYNA-N
1,7,7-trimethylbicyclo[2.2.1]heptan-2-one
CC1(C)C2CCC1(C)C(=O)C2

CA185, 76-22-2, C10H16O

Spectrum™ Chemical Camphor, Synthetic, USP is used as a decongestant, antiseptic stimulant and anesthetic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN2717
Quantity 110 lb.
Grade USP
Packaging Fiber Drum
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit