Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ cAMPS-Rp, triethylammonium salt
GREENER_CHOICESDP

Catalog No. 13371 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-371 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-371 Supplier Tocris Bioscience™ Supplier No. 1337/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

cAMP antagonist

Cell-permeable cAMP analog; acts as a competitive antagonist of cAMP-induced activation of PKA (IC50 = 11 - 16 μM) by interacting with cAMP binding sites on the regulatory subunits. Resistant to hydrolysis by phosphodiesterases.

Chemical Identifiers

CAS 151837-09-1
Molecular Formula C16H27N6O5PS

Specifications

Quantity 1 mg
Formula Weight 446.46
Purity >98%
Molecular Weight (g/mol) 446.463
Chemical Name or Material cAMPS-Rp, triethylammonium salt
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.