Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Camptothecin 97% MP Biomedicals

Catalog No. p-3178702
Click to view available options
Quantity:
25 mg
50 mg
250 mg

Chemical Identifiers

CAS 7689-03-4
Molecular Formula C20H16N2O4
Molecular Weight (g/mol) 348.36
MDL Number MFCD00081076
InChI Key VSJKWCGYPAHWDS-FQEVSTJZSA-N
Synonym camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecin
PubChem CID 24360
ChEBI CHEBI:27656
SMILES CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O

Specifications

Color Yellow
Density 1.51g/cm3
Assay Percent Range >97%
Quantity 25 mg
Formula Weight 348.3
Physical Form Solid
Chemical Name or Material Camptothecin
Recommended Storage Store at + 4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.