Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Camptothecin
GREENER_CHOICESDP

Catalog No. 110025 Shop All Novus Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-002-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-002-5 Supplier R&D Systems™ Supplier No. 1100/25
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. DNA topoisomerase inhibitor

Cytotoxic plant alkaloid with antitumor properties; prototypic DNA topoisomerase I inhibitor. Induces single strand DNA breaks and protein-DNA crosslinks.

Chemical Identifiers

CAS 3-4-7689
Molecular Formula C20H16N2O4

Specifications

Quantity 25 mg
Product Line Tocris
Formula Weight 348.36
Purity >98%
Molecular Weight (g/mol) 348.36
Chemical Name or Material Camptothecin
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.