Learn More
3M™ Campylobacter Enrichment Broth CE250, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26759 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Campylobacter Enrichment Broth CE250, Sold by AquaPhoenix Scientific
Catalog No.
S26759 Campylobacter Enrichment Broth CE250, Sold by AquaPhoenix Scientific
1 options
Catalog No. S26759 Supplier 3M™ Supplier No. CE250
Aerobically incubated medium used for selective enrichment of Campylobacter
  • Aerobic incubation
    • No need for microaerophilic environment
  • Ready to use enrichment
    • Just add sterile water (no supplements or organic solvents)
  • Use with MDA2CAM96 3M™ molecular detection assay 2
    • Campylobacter for fast and reliable results
  • 250 g bottle makes approximately 203 bird rinse enrichments
  • Real-time amplification and detection process identifies positive results in as early as 15 mi.
    • For making critical decisions fast
Specifications
Product Type Microbiology Media
For Use With (Application) Selective enrichment of Campylobacter
For Use With (Equipment) MDA2CAM96 3M™ molecular detection assay 2
Weight (Metric) 250 g