Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Campylobacter Jejuni Antibody (380/412), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062426AF594
Description
Campylobacter Jejuni Monoclonal specifically detects Campylobacter Jejuni in Bacteria samples. It is validated for ELISA.Specifications
Campylobacter Jejuni | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse | |
0.1 mL | |
NB100-62426 is specific for Campylobacter jejuni. Some reactivity has been observed with Penner 4 +/- and with Penner 10. C. jejuni is a curved, rod-shaped gram negative bacterium which is a member of the Campylobacteraceae family. They are motile with a polar unsheathed flagellum that results in a corkscrew like motion. They occur primarily as commensals in humans and domestic animals, particularly in chickens where gut temperatures of 42oC are optimal for growth. C. jejuni are one of the most common causes of human diarrhea in the world. Food poisoning can be severely debilitating, although rarely life-threatening as the disease is self limiting. Campylobacter infections are also associated with postinfectious complications including arthritis, Reiter syndrome and GuillainûBarre┤ syndrome. | |
Store at 4°C in the dark. |
ELISA | |
380/412 | |
ELISA | |
Campilobacter | |
C. jejuni Penner 2 | |
Primary | |
Bacteria | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction