Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Campylobacter Jejuni Antibody (380/412) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Campylobacter Jejuni |
---|---|
Clone | 380/412 |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1:100-1:2000 |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017472
![]() |
Novus Biologicals
NB100624260.025MG |
0.025 mg |
Each for $206.00
|
|
|||||
NB10062426
![]() |
Novus Biologicals
NB10062426 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
Campylobacter Jejuni Monoclonal specifically detects Campylobacter Jejuni in Bacteria samples. It is validated for ELISA.Specifications
Campylobacter Jejuni | |
1.0 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Bacteria | |
C. jejuni Penner 2 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
380/412 | |
ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Campilobacter | |
IgG2a | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title