Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Campylobacter jejuni Monoclonal Antibody (J92D)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA518175
Description
This antibody does not cross-react with H. pylori, E. coli (several), Salmonellae (several), Shigellae (several), P. aeruginosa, Yersinia or Citrobacter. Purity is >90%.
Campylobacter jejuni is a species of curved, long rod-shaped, Gram-negative microaerophilic, bacteria commonly found in animal feces. It is one of the most common causes of human gastroenteritis in the world. C. jejuni is commonly associated with poultry and naturally colonizes the G1 tract of many bird species.
Specifications
Campylobacter jejuni | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG2a |
ELISA, Western Blot | |
J92D | |
Unconjugated | |
C. jejuni | |
Campylobacter jejuni native protein. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction