Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Campylobacter jejuni Monoclonal Antibody (J99F)
GREENER_CHOICE

Catalog No. PIMA17473
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17473 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17473 Supplier Invitrogen™ Supplier No. MA17473
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7473 antibody reacts with Campylobacter jejuni 100 kDa in bacterial and infected tissue samples. MA1-7473 has been successfully used in WB applications. The MA1-7473 antibody was raised against the C. jejuni 100 kDa antigen.

Campylobacter jejuni is a species of curved, long rod-shaped, Gram-negative microaerophilic, bacteria commonly found in animal feces. It is one of the most common causes of human gastroenteritis in the world. C. jejuni is commonly associated with poultry and naturally colonizes the G1 tract of many bird species.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Campylobacter jejuni
Applications Western Blot
Classification Monoclonal
Clone J99F
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias C. jejuni
Host Species Mouse
Immunogen 100 kDa antigen.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.