Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Campylobacter Jejuni Antibody (BGN/2E-10), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | Campylobacter Jejuni |
|---|---|
| Clone | BGN/2E-10 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), ELISA, Immunofluorescence |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017684
![]() |
Novus Biologicals
NB100650390.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10065039
![]() |
Novus Biologicals
NB10065039 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Campylobacter Jejuni Monoclonal specifically detects Campylobacter Jejuni in Bacteria samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
| Campylobacter Jejuni | |
| 1 mg/mL | |
| Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), ELISA, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| Native, Type 1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| BGN/2E-10 | |
| ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| Campilobacter | |
| IgM | |
| Ammonium sulfate precipitation | |
| NB100-65039 reacts with C. jejuni type 1 (K807, K858, K634), C. coli, C. hyointestinalis, C. lardis, C. jejuni type 2. It cross reacts with S. aureus and weakly with P. fluorescens. NB100-65039 reacts with a soluble excreted antigen in EIA. This antibody is unaffected by frozen storage of specimens, unlike antibodies to flagellar antigens which require fresh cultured organisms. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title