Learn More
Invitrogen™ Campylobacter jejuni Polyclonal Antibody

Description
PA1-7205 detects Campylobacter jejuni. PA1-7205 has been successfully used in ELISA procedures. The PA1-7205 immunogen is ATCC strain 29428.
Specifications
Specifications
| Antigen | Campylobacter jejuni |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | C. jejuni |
| Host Species | Rabbit |
| Immunogen | Campylobacter jejuni whole cells. |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.