Learn More
Invitrogen™ CAMSAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116776
Description
Positive Samples: HeLa Immunogen sequence: KTVLHQKSSR PPVPISNATK RSFLGSPAAG TLAELQPPVQ LPAEGCHRHY LHPEEPEYLG KGTAAFSPSH PLLPLRQKQQ KSIQGEDIPD QRHRSNSLTR VDGQPRGAAI AWPEKKTRPA SQPTPFALHH AASCEVDPSS GDSISLARSI SKDSLASNIV NLTPQNQPHP TATKSHGKSL LSNVSIEDEE EELVAIVRAD VVPQQADPEF PRASPRALGL TANARSPQGQ LDTSESKPDS FFLEPLMPAV LKPAKEKQVI TKEDERGEGR PRSIVSRRPS EGPQPLVRRK MTGSRDLNRT F.
Key microtubule-organizing protein that specifically binds the minus-end of non-centrosomal microtubules and regulates their dynamics and organization (PubMed:19508979, PubMed:21834987, PubMed:24486153, PubMed:24706919, PubMed:24117850). Specifically recognizes growing microtubule minus-ends and stabilizes microtubules (PubMed:24486153, PubMed:24706919). Acts on free microtubule minus-ends that are not capped by microtubule-nucleating proteins or other factors and protects microtubule minus-ends from depolymerization (PubMed:24486153, PubMed:24706919). In contrast to CAMSAP2 and CAMSAP3, tracks along the growing tips of minus-end microtubules without significantly affecting the polymerization rate: binds at the very tip of the microtubules minus-end and acts as a minus-end tracking protein (-TIP) that dissociates from microtubules after allowing tubulin incorporation (PubMed:24486153, PubMed:24706919). Through interaction with spectrin may regulate neurite outgrowth (PubMed:24117850). [UniProt].
Specifications
| CAMSAP1 | |
| Polyclonal | |
| Unconjugated | |
| CAMSAP1 | |
| 9530003A05Rik; C77823; calmodulin regulated spectrin associated protein 1; calmodulin regulated spectrin-associated protein 1; calmodulin-regulated spectrin-associated protein 1; Camsap1; DKFZp434F195; FLJ31228; PRO2405; RGD1565022 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 157922, 227634 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.83 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| A2AHC3, Q5T5Y3 | |
| CAMSAP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 350-650 of human CAMSAP1 (NP_0562623). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.