Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMSAP1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB166201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166201 100 μg
NB166202 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166201 Supplier Novus Biologicals Supplier No. NBP315553100UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CAMSAP1 Polyclonal antibody specifically detects CAMSAP1 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen CAMSAP1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias bA100C15.1, bA100C15.1 (novel protein), calmodulin regulated spectrin-associated protein 1, calmodulin-regulated spectrin-associated protein 1, DKFZp434F195, DKFZp434G2311, FLJ31228, MGC163452, PRO2405
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-650 of human CAMSAP1 (NP_056262.3).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 157922
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.