Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMSAP1L1 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP121401AF594
Description
CAMSAP1L1 Polyclonal antibody specifically detects CAMSAP1L1 in Human samples. It is validated for Western BlotSpecifications
CAMSAP1L1 | |
Polyclonal | |
Western Blot | |
calmodulin regulated spectrin-associated protein 1-like 1, calmodulin-regulated spectrin-associated protein 2, CAMSAP2, KIAA1078Calmodulin-regulated spectrin-associated protein 1-like protein 1, MGC150680, MGC150681 | |
The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human calmodulin regulated spectrin-associated protein 1-like 1 using the numbering given in entry EAW91320.1 (GeneID 23271). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23271 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction