Learn More
CAMSAP1L1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302161A

Description
The recommended shelf life for this product is 1 year from date of receipt.
CAMSAP2 gene ontology annotations related to this gene include neuron projection development.Specifications
CAMSAP1L1 | |
Polyclonal | |
Unconjugated | |
CAMSAP2 | |
calmodulin regulated spectrin-associated protein 1-like 1, calmodulin regulated spectrin-associated protein family, member 2, calmodulin-regulated spectrin-associated protein 1-like protein 1, CAMSAP1L1, KIAA1078 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23271 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q08AD1 | |
CAMSAP2 | |
Between 375 and 425 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.