Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMSAP2 Mouse anti-Human, Mouse, Rat, Clone: 2A10A1, Proteintech
SDP

Catalog No. 501736888 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6888 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6888 Supplier Proteintech Group Inc Supplier No. 664631IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CAMSAP2 gene ontology annotations related to this gene include neuron projection development.

Specifications

Antigen CAMSAP2
Applications Western Blot
Classification Monoclonal
Clone 2A10A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CAMSAP2
Gene Accession No. Q08AD1, Q8C1B1
Gene Alias CAMSAP1L1, CAMSAP2, KIAA1078
Gene Symbols Camsap2
Host Species Mouse
Immunogen CAMSAP2 Fusion Protein Ag11794
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23271, 289400, 67886
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.