Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAMSAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555301 Supplier Invitrogen™ Supplier No. PA555301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSVHGVSFDI SFDKEDSVQR STPNRGITRS ISNEGLTLNN SHVSKHIRKN LSFKPINGEE EAESIEEELN IDSHSDLKSC VPL Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 77%.

CAMSAP2 gene ontology annotations related to this gene include neuron projection development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAMSAP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAMSAP2
Gene Accession No. Q08AD1
Gene Alias 1600013L13Rik; 4930541M15Rik; calmodulin regulated spectrin associated protein family member 2; calmodulin regulated spectrin-associated protein 1-like 1; calmodulin regulated spectrin-associated protein family, member 2; calmodulin-regulated spectrin-associated protein 1-like protein 1; Calmodulin-regulated spectrin-associated protein 2; CAMSAP1L1; CAMSAP2; KIAA1078; mKIAA1078; RGD1310950
Gene Symbols CAMSAP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CAMSAP2. Recombinant protein control fragment (Product #RP-94138).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23271
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.