Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAMSAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560528 Supplier Invitrogen™ Supplier No. PA560528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRHQPILMGA HLAVIDALMA AFAFEWTKTL PGPLALTSLE HKLLFWVDTT VRRLQEKTEQ EAAQRASPAA PADGAAPAQP SIRYRKDR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

Key microtubule-organizing protein that specifically binds the minus-end of non-centrosomal microtubules and regulates their dynamics and organization (PubMed:19041755, PubMed:23169647). Specifically recognizes growing microtubule minus-ends and autonomously decorates and stabilizes microtubule lattice formed by microtubule minus-end polymerization (PubMed:24486153). Acts on free microtubule minus-ends that are not capped by microtubule-nucleating proteins or other factors and protects microtubule minus-ends from depolymerization (PubMed:24486153). In addition, it also reduces the velocity of microtubule polymerization (PubMed:24486153). Required for the biogenesis and the maintenance of zonula adherens by anchoring the minus-end of microtubules to zonula adherens and by recruiting the kinesin KIFC3 to those junctional sites (PubMed:19041755). Required for orienting the apical-to-basal polarity of microtubules in epithelial cells: acts by tethering non-centrosomal microtubules to the apical cortex, leading to their longitudinal orientation (PubMed:27802168, PubMed:26715742). Plays a key role in early embryos, which lack centrosomes: accumulates at the microtubule bridges that connect pairs of cells and enables the formation of a non-centrosomal microtubule-organizing center that directs intracellular transport in the early embryo. Couples non-centrosomal microtubules with actin: interaction with MACF1 at the minus ends of non-centrosomal microtubules, tethers the microtubules to actin filaments, regulating focal adhesion size and cell migration (PubMed:27693509). Plays a key role in the generation of non-centrosomal microtubules by accumulating in the pericentrosomal region and cooperating with KATNA1 to release non-centrosomal microtubules from the centrosome (PubMed:28386021). Through the microtubule cytoskeleton, also regulates the organization of cellular organelles including the Golgi and the early endosomes (PubMed:28089391). Through interaction with AKAP9, involved in translocation of Golgi vesicles in epithelial cells, where microtubules are mainly non-centrosomal (PubMed:28089391). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAMSAP3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CAMSAP3
Gene Accession No. Q9P1Y5
Gene Alias 2310057J16Rik; calmodulin regulated spectrin associated protein family member 3; calmodulin regulated spectrin-associated protein family, member 3; calmodulin-regulated spectrin-associated protein 3; CAMSAP3; KIAA1543; Marshalin; marshalin-La; marshalin-Lb; marshalin-Lc; marshalin-Ld; marshalin-Sa; marshalin-Sb; marshalin-Sc; marshalin-Sd; Nezha; PPP1R80; Protein Nezha; protein phosphatase 1, regulatory subunit 80
Gene Symbols CAMSAP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CAMSAP3. Recombinant protein control fragment (Product #RP-98884).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.