Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAMTA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578440
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578440 Supplier Invitrogen™ Supplier No. PA578440
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: human CAMTA1-transfected 293T Predicted Reactivity: Mouse (88%), Rat (91%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CAMTA1 is a transcriptional activator. May act as a tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAMTA1
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene CAMTA1
Gene Accession No. Q9Y6Y1
Gene Alias 1810059M14Rik; 2310058O09Rik; AI316882; AW987475; calmodulin binding transcription activator 1; calmodulin-binding transcription activator 1; Camta1; CANPMR; Kiaa0833; mKIAA0833; MSTP023
Gene Symbols CAMTA1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CAMTA1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23261
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.