Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CAMTA1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA557619
Description
Immunogen sequence: ESLSMLPTNV SEELVLSTTL DGGRKIPETT MNFDPDCFLN NPKQGQTYGG GGLKAEMVSS NIRHSPPGER SFSFTTVLTK EIKTE Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
Transcriptional activator. May act as a tumor suppressor.
Specifications
CAMTA1 | |
Polyclonal | |
Unconjugated | |
CAMTA1 | |
calmodulin binding transcription activator 1, CANPMR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23261 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6Y1 | |
CAMTA1 | |
Recombit protein corresponding to Human CAMTA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction