Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CAMTA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568124
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568124 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568124 Supplier Invitrogen™ Supplier No. PA568124
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

Transcription activator. May act as tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CAMTA2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CAMTA2
Gene Accession No. O94983, Q80Y50
Gene Alias calmodulin binding transcription activator 2; calmodulin-binding transcription activator 2; Camta2; Kiaa0909; Kiaa0909-hp; mKIAA0909
Gene Symbols CAMTA2
Host Species Rabbit
Immunogen Synthesized peptide derived from human CAMTA2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216874, 23125
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.