Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CAMTA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5140051
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LPPASDDGAA PEDADSPQAV DVIPVDMISL AKQIIEATPE RIKREDFVGL PEAGASMRER TGAVGLSETM SWLASYLENV DHFPSSTPPS ELPFERGRLA VPSAPSWAEF.
Transcription activator. May act as tumor suppressor.
Specifications
| CAMTA2 | |
| Polyclonal | |
| Unconjugated | |
| CAMTA2 | |
| calmodulin binding transcription activator 2; calmodulin-binding transcription activator 2; Camta2; Kiaa0909; Kiaa0909-hp; mKIAA0909 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23125 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94983 | |
| CAMTA2 | |
| Recombinant protein corresponding to Human CAMTA2. Recombinant protein control fragment (Product #RP-108774). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction