Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cancer Diagnostics, Inc.™ Tissue Marking Dyes
SDP

Catalog No. 22500021 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Quantity:
2 oz.
8 oz.
Color:
Black
Blue
Green
Green, Black, Red, Yellow, Blue, Orange
Green, Black, Red, Yellow, Blue, Orange, Violet
Orange
Red
Yellow
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
22-500-021 8 oz. Red
22-500-017 2 oz. Orange
22-500-014 2 oz. Green
22-500-031 2 oz. Green, Black, Red, Yellow, Blue, Orange
22-500-466 2 oz. Green, Black, Red, Yellow, Blue, Orange, Violet
22-500-012 2 oz. Black
22-500-013 2 oz. Blue
22-500-015 2 oz. Red
22-500-019 8 oz. Blue
22-500-016 2 oz. Yellow
22-500-018 8 oz. Black
22-500-022 8 oz. Yellow
22-500-020 8 oz. Green
22-500-023 8 oz. Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 22-500-021 Supplier Cancer Diagnostics, Inc.™ Supplier No. 07284
Add to Cart
Add to Cart

Formulated to be used without a mordant

  • Dyes are pigment-based with an alcohol foundation for consistency and durability
  • Proprietary blend of materials remains bright through processing and produces clean, crisp results
  • Will not run or bleed on tissue
  • Impervious to laboratory chemicals and reagents

Order Info

Individual colors available in two sizes; also available in systems with sets of six or seven colors, wood tray and 50 applicator sticks. Two-year shelf life.

Specifications

Boiling Point 93°C
Color Red
Product Type Tissue Marking Dye
Quantity 8 oz.
Formulation Isopropyl Alcohol 400ppm, Ammonia 35ppm
pH 8.5 to 10
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.