Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cancer Diagnostics, Inc.™ Bio Board
SDP

Catalog No. 22500462 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Dimensions (L x W):
8 x 10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-500-462 8 x 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-500-462 Supplier Cancer Diagnostics, Inc.™ Supplier No. BB081160
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for routine grossing, frozen sections, and photography

  • Cut-resistant, disposable foam core board
  • Light blue color provides exceptional color contrast
  • Can be used for multiple cases—use different quadrants and wipe off

Specifications

Product Type Bio Board™
Color Light Blue
For Use With (Application) For routine grossing, frozen sections, and photography
Dimensions (L x W) 8 x 10 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.