Learn More
Cancer Diagnostics, Inc.™ Cytology Staining Reagents
SDP
Catalog No. 22500047 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Product Type:
EA-50 Cytology Counter Stain
EA-50 Modified Cytology Counter Stain
EA-65 Cytology Counter Stain
OG-6 Cytology Stain
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-500-047 EA-50 Modified Cytology Counter Stain
22-500-046 EA-50 Cytology Counter Stain
22-500-048 EA-65 Cytology Counter Stain
22-500-049 OG-6 Cytology Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-500-047 Supplier Cancer Diagnostics, Inc.™ Supplier No. CM8954
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use in routine cytoplasmic staining
EA-50, EA-50 Modified, and EA-65 are alcohol-based stains used in the cytology laboratory for routine cytoplasmic staining using thePapanicolaou Method.
Orange G-6 is an alcohol-based stain used in the cytology laboratory to detect the presence of keratin in the cytoplasm. This formula is pre filtered before bottling and is manufactured for optimal consistency.
For routine cytoplasmic staining using Papanicolaou Method
Specifications
For Use With (Application) For routine cytoplasmic staining using papanicolaou method
Quantity 3800mL
Product Type EA-50 Modified Cytology Counter Stain