Description
EA-50, EA-50 Modified, and EA-65 are alcohol-based stains used in the cytology laboratory for routine cytoplasmic staining using thePapanicolaou Method.
Orange G-6 is an alcohol-based stain used in the cytology laboratory to detect the presence of keratin in the cytoplasm. This formula is pre filtered before bottling and is manufactured for optimal consistency.
For routine cytoplasmic staining using Papanicolaou Method
Specifications
Specifications
| For Use With (Application) | For routine cytoplasmic staining using papanicolaou method |
| Quantity | 3800mL |
| Product Type | EA-65 Cytology Counter Stain |