Description
All stains are filtered prior to bottling and use a special process to ensure optimal freshness.
Mercury Free Harris Hematoxylin produces a deep, dark blue-purple stain. It is generally used in regressive methods, but may also be used progressively both in histology and cytology preparations.
Gill 1,2,3 generally used in the progressive method.
Mayer's Hematoxylin used for routine H&E and immunochemistry.
Used in progressive method for histology and cytology preparations
Specifications
Specifications
| For Use With (Application) | Used in progressive method for histology and cytology preparations |
| Quantity | 3800mL |
| Product Type | Gill 1 Hematoxylin |