Learn More
Cancer Diagnostics, Inc.™ Hinged Tissue Cassettes (Slat Holes) - Taped Bases and Lids
Shop All Cancer Diagnostics Products
Description
- Optimal printing and writing surface
- Available in 11 colors
- Taped bases and lids are packed separately
Specifications
Specifications
| Pore Style | Slat Holes |
| Closure Type | Taped Bases and Lid |
| Color | Yellow |
| Product Type | Tissue Cassette |