Learn More
Cancer Diagnostics, Inc.™ Hinged Tissue Cassettes (Slat Holes) - Taped Bases and Lids
SDP
Catalog No. 22500645 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Orange
Peach
Tan
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-500-645 Aqua
22-500-636 White
22-500-639 Green
22-500-637 Gray
22-500-638 Blue
22-500-641 Yellow
22-500-642 Orange
22-500-646 Tan
22-500-644 Peach
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-500-645 Supplier Cancer Diagnostics, Inc.™ Supplier No. SHLTP1009
Add to Cart
Add to Cart
This item is not returnable. View return policy
Hinged slat-style tissue cassettes
  • Optimal printing and writing surface
  • Available in 11 colors
  • Taped bases and lids are packed separately
Specifications
Pore Style Slat Holes
Closure Type Taped Bases and Lid
Color Aqua
Product Type Tissue Cassette