Learn More
Cancer Diagnostics, Inc.™ Molded 1-Piece Tissue Cassettes (Slat Holes) - Taped Stacks
SDP
Catalog No. 22500620 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lavender
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-500-620 Peach
22-500-613 Gray
22-500-614 Blue
22-500-616 Pink
22-500-612 White
22-500-617 Yellow
22-500-615 Green
22-500-618 Orange
22-500-619 Lavender
22-500-621 Aqua
22-500-622 Tan
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 22-500-620 Supplier Cancer Diagnostics, Inc.™ Supplier No. SCTP1008
Add to Cart
Add to Cart
Optimal polymer technology resistant to all chemicals
  • Optimal writing surface
  • Excellent adherence for all inks
  • Available in 11 colors
Specifications
Pore Style Slat Holes
Color Peach
Product Type Tissue Cassette