Learn More
Cancer Diagnostics, Inc.™ Molded Biopsy Micro Cassettes (Mesh) - Taped
SDP
Catalog No. 22500681 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lavender
Peach
Pink
Tan
White
Yellow
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-500-681 Gray
22-500-680 White
22-500-682 Blue
22-500-683 Green
22-500-684 Pink
22-500-685 Yellow
22-500-687 Lavender
22-500-690 Tan
22-500-689 Aqua
22-500-688 Peach
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 22-500-681 Supplier Cancer Diagnostics, Inc.™ Supplier No. MHT1001
Add to Cart
Add to Cart
Molded mesh for ultimate biopsy security
  • Molded for optimal surface tension
  • Allows for complete processing
  • Package in taped stacks of 2000 per case
Specifications
Pore Style Mesh
Color Gray
Product Type Biopsy Cassette