Learn More
Cancer Diagnostics, Inc.™ Molded Biopsy Micro Cassettes (Mesh) - Taped
Shop All Cancer Diagnostics Products
Description
- Molded for optimal surface tension
- Allows for complete processing
- Package in taped stacks of 2000 per case
Specifications
Specifications
| Pore Style | Mesh |
| Color | Peach |
| Product Type | Biopsy Cassette |