Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cancer Diagnostics, Inc.™ Nylon Mesh Biopsy Bags
SDP

Catalog No. p-3198932
Click to view available options
Color:
White
Yellow
Quantity:
Pack of 100
Pack of 1000

Reduce cross-contamination

  • Made from nylon mesh with precise mesh diameter
  • Allow tiny specimens to be captured by straining the fixative when poured into the bag
  • Can be used to contain tiny biopsies during decalcification
  • Simply insert specimen, fold bag to enclose cassette or capsule, and process

Order Info

Measure 30 x 45mm; available in white and yellow.

Specifications

Color White
Dimensions (L x W) 30 x 40 mm
Quantity Pack of 1000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.