Learn More
Cancer Diagnostics, Inc.™ Processing Cassettes (Round Hole) - Taped
SDP
Catalog No. 22500629 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Orange
Peach
Pink
Tan
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-500-629 Orange
22-500-627 Pink
22-500-626 Green
22-500-625 Blue
22-500-624 Gray
22-500-628 Yellow
22-500-633 Tan
22-500-632 Aqua
22-500-631 Peach
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-500-629 Supplier Cancer Diagnostics, Inc.™ Supplier No. LHC1006T
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed for use with all industry metal lids
  • Lids not included (metal and plastic lids available separately)
  • 11 different stock colors available
Specifications
Pore Style Round Holes
Closure Type Separate Lid
Color Orange
Product Type Processing Cassette