Description
- Rounded interiors without corners
- One-piece breakable lid design
- Filtration enhanced by side vents
- Processed biopsies are easy to find
- Small and assorted colors available
- Loose and taped options for packaging
Specifications
Specifications
| Packaging Type | Loose |
| Color | Green |
| Product Type | Vortex Biopsy Cassette |