Learn More
Cancer Diagnostics, Inc.™ Vortex™ Biopsy Cassettes
SDP
Catalog No. 22500889 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
Packaging Type:
Loose
Taped
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Packaging Type
22-500-889 Yellow Loose
22-500-885 White Loose
22-500-887 Green Loose
22-500-890 White Taped
22-500-888 Pink Loose
22-500-891 Blue Taped
22-500-893 Pink Taped
22-500-892 Green Taped
22-500-894 Yellow Taped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-500-889 Supplier Cancer Diagnostics, Inc.™ Supplier No. VB1005
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for routine processing and embedding of tissues and cells during histological studies
  • Rounded interiors without corners
  • One-piece breakable lid design
  • Filtration enhanced by side vents
  • Processed biopsies are easy to find
  • Small and assorted colors available
  • Loose and taped options for packaging
Specifications
Packaging Type Loose
Color Yellow
Product Type Vortex Biopsy Cassette