Learn More
Cancer Diagnostics, Inc.™ Vortex™ Biopsy Micro Cassettes
SDP
Catalog No. 22500875 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Color:
Blue
Green
White
Yellow
Packaging Type:
Loose
Taped
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Packaging Type
22-500-875 White Taped
22-500-871 Blue Loose
22-500-870 White Loose
22-500-872 Green Loose
22-500-874 Yellow Loose
22-500-876 Blue Taped
22-500-877 Green Taped
22-500-879 Yellow Taped
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-500-875 Supplier Cancer Diagnostics, Inc.™ Supplier No. VMT1000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Used for routine processing and embedding of tissues and cells during histological studies
  • Rounded micro hole interior basket without corners
  • Filtration enhanced by side vents
  • Two-piece lid design
  • Processed biopsies are easy to find
  • Various colors available
  • Tubed, loose and taped options for packaging
Specifications
Packaging Type Taped
Color White
Product Type Biopsy Micro Cassette